Potassium(2-fluorophenyl)trifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(2-fluorophenyl)borate, (2-Fluorophenyl)trifluoroboric acid potassium salt

A289482-1G A289482-250MG A289482-5G
77014-748EA 46 USD
77014-748 77014-750 77014-752
Potassium(2-fluorophenyl)trifluoroborate 98%
Potassium(2-fluorophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₆H₄BF₄K
MW: 202
MDL Number: MFCD06659937
CAS Number: 166328-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR