2-Bromo-4-methyl-5-nitrobenzaldehyde 95%

Supplier: Ambeed
Warning

A289426-1G A289426-250MG A289426-5G
76988-624EA 75 USD
76988-624 76988-626 77890-536
2-Bromo-4-methyl-5-nitrobenzaldehyde 95%
2-Bromo-4-methyl-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrNO₃
MDL Number: MFCD13185992
CAS Number: 159730-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR