6-Chloro-2-methylpyrido[3,4-d]pyrimidin-4(1H)-one 97%

Supplier: Ambeed
Warning

A289410-100MG A289410-250MG A289410-5G
77014-742EA 47 USD
77014-742 77014-744 77014-746
6-Chloro-2-methylpyrido[3,4-d]pyrimidin-4(1H)-one 97%
6-Chloro-2-methylpyrido[3,4-d]pyrimidin-4(1H)-one

Precaución: for Research Use Only

Formula: C₈H₆ClN₃O
MDL Number: MFCD27996796
CAS Number: 1637671-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR