Methyl-4-fluoro-3-methylbenzoate 98%

Supplier: Ambeed
Warning

A289388-100G A289388-10G A289388-1G A289388-25G A289388-5G
77002-332EA 283 USD
77002-332 77002-334 77002-336 77002-338 77002-340
Methyl-4-fluoro-3-methylbenzoate 98%
Methyl 4-fluoro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MDL Number: MFCD06203783
CAS Number: 180636-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR