Tetradecyltrimethylammonium bromide 98%

Supplier: Ambeed
Warning

Synonyms: MiTMAB, (1-Tetradecyl)trimethylammonium bromide, N,N,N-Trimethyltetradecan-1-aminium bromide, Tetradonium bromide, Tetradecyl trimethyl ammonium Bromide, Myristyltrimethylammonium bromide, Dynamin Inhibitor II, TTAB

A289209-1000G A289209-100G A289209-25G A289209-500G
77002-320EA 78 USD
77002-320 77002-322 77002-324 77002-326
Tetradecyltrimethylammonium bromide 98%
Tetradecyltrimethylammonium bromide

Precaución: for Research Use Only

Formula: C₁₇H₃₈BrN
MW: 336.394 g/mol
MDL Number: MFCD00011770
CAS Number: 1119-97-7
Merck Index: 12,06419

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR