tert-Butyl ((1H-indol-5-yl)methyl)carbamate 95%

Supplier: Ambeed
Warning

A289121-100MG A289121-250MG
76988-620EA 38 USD
76988-620 76988-622
tert-Butyl ((1H-indol-5-yl)methyl)carbamate 95%
tert-Butyl ((1H-indol-5-yl)methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₂
MDL Number: MFCD06656839
CAS Number: 267875-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR