3-Bromo-9-(2-naphthyl)carbazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-9-(naphthalen-2-yl)-9H-carbazole
A289100-100G
A289100-1G
A289100-25G
A289100-5G
77014-726EA
512.42
USD
77014-726
77014-728
77014-730
77014-732
3-Bromo-9-(2-naphthyl)carbazole 97%
3-Bromo-9-(2-naphthyl)carbazole
Precaución: for Research Use Only
Formula:
C₂₂H₁₄BrN MW: 372.27 g/mol |
MDL Number:
MFCD26127422 CAS Number: 934545-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...