3-(Dimethylamino)benzhydrazide 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Dimethylamino)benzohydrazide

A289044-1G A289044-250MG
77032-270EA 63.66 USD
77032-270 77032-272
3-(Dimethylamino)benzhydrazide 98%
3-(Dimethylamino)benzhydrazide

Precaución: for Research Use Only

Formula: C₉H₁₃N₃O
MW: 179.2190 g/mol
MDL Number: MFCD00238588
CAS Number: 205927-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR