Ethyl-5-amino-1-(4-chlorophenyl)-1H-pyrazole-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-amino-1-(4-chlorophenyl)-1H-pyrazole-4-carboxylate

A288962-1G A288962-250MG A288962-25G
77002-316EA 23.34 USD
77002-316 77002-318 77672-486
Ethyl-5-amino-1-(4-chlorophenyl)-1H-pyrazole-4-carboxylate 95%
Ethyl-5-amino-1-(4-chlorophenyl)-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClN₃O₂
MW: 265.7 g/mol
MDL Number: MFCD00973905
CAS Number: 14678-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR