3-(Methylthio)phenylboronic acid pinacolate 95%

Supplier: Ambeed
Warning

Synonyms: 3-Methylthiophenylboronic acid pinacol ester, 4,4,5,5-Tetramethyl-2-(3-(methylthio)phenyl)-1,3,2-dioxaborolane

A288952-10G A288952-1G A288952-250MG A288952-5G
77032-254EA 48 USD
77032-254 77032-256 77032-258 77032-260
3-(Methylthio)phenylboronic acid pinacolate 95%
3-(Methylthio)phenylboronic acid pinacolate

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₂S
MDL Number: MFCD05155220
CAS Number: 710348-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR