Ethyl 2-(trifluoromethyl)pyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 5-(Ethoxycarbonyl)-2-(trifluoromethyl)pyrimidine

A288598-10G A288598-1G A288598-250MG A288598-5G
77032-246EA 206.88 USD
77032-246 77032-248 77032-250 77032-252
Ethyl 2-(trifluoromethyl)pyrimidine-5-carboxylate 97%
Ethyl 2-(trifluoromethyl)pyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₇F₃N₂O₂
MW: 220.149 g/mol
MDL Number: MFCD09754048
CAS Number: 304693-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR