Methyl-4-amino-2-chloropyrimidine-5-carboxylate 95%

Supplier: Ambeed
Warning

A288586-1G A288586-250MG
77002-312EA 84.01 USD
77002-312 77002-314
Methyl-4-amino-2-chloropyrimidine-5-carboxylate 95%
Methyl-4-amino-2-chloropyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆ClN₃O₂
MDL Number: MFCD14585039
CAS Number: 858269-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR