Fmoc-O-t-butyl-L-β-homoserine 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-β-HomoSer(tBu)-OH, Fmoc-β-HoSer(tBu)-OH

A288572-100MG A288572-1G A288572-250MG A288572-5G
77002-302EA 60 USD
77002-302 77002-306 77002-308 77002-310
Fmoc-O-t-butyl-L-β-homoserine 95%
Fmoc-O-t-butyl-L-beta-homoserine

Precaución: for Research Use Only

Formula: C₂₃H₂₇NO₅
MDL Number: MFCD01862865
CAS Number: 203854-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR