Methyl 2-(5-bromopyridin-2-yl)acetate 98%

Supplier: Ambeed
Warning

A288245-10G A288245-1G A288245-250MG A288245-25G A288245-5G
77014-714EA 449.83 USD
77014-714 77014-716 77014-718 77014-720 77014-722
Methyl 2-(5-bromopyridin-2-yl)acetate 98%
Methyl 2-(5-bromopyridin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.05 g/mol
MDL Number: MFCD12031821
CAS Number: 917023-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR