3-Chloro-4-(methylsulfonyl)-1-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-(methylsulphonyl)nitrobenzene

A288140-1G A288140-250MG A288140-5G
77002-294EA 47.75 USD
77002-294 77002-296 77002-298
3-Chloro-4-(methylsulfonyl)-1-nitrobenzene 98%
3-Chloro-4-(methylsulfonyl)-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₆ClNO₄S
MW: 235.65 g/mol
MDL Number: MFCD08692409
CAS Number: 91842-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR