2-Chloro-4-fluoro-5-iodobenzoic acid 95%

Supplier: Ambeed
Warning

A288080-100MG A288080-250MG
76988-612EA 10.61 USD
76988-612 76988-614
2-Chloro-4-fluoro-5-iodobenzoic acid 95%
2-Chloro-4-fluoro-5-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃ClFIO₂
MW: 300.45 g/mol
MDL Number: MFCD24107204
CAS Number: 264927-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR