5-Bromo-2-methyl-4-nitropyridine-N-oxide 95%
Supplier: Ambeed
|
Warning
|
A287949-100MG
A287949-1G
A287949-250MG
A287949-5G
76988-602EA
18
USD
76988-602
76988-604
76988-606
76988-608
5-Bromo-2-methyl-4-nitropyridine-N-oxide 95%
5-Bromo-2-methyl-4-nitropyridine-N-oxide
Precaución: for Research Use Only
Formula:
C₆H₅BrN₂O₃ |
MDL Number:
MFCD11977426 CAS Number: 62516-08-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...