(S)-Benzyl 4-isopropyl-2,5-dioxooxazolidine-3-carboxylate 95%

Supplier: Ambeed
Warning

A287141-100G A287141-1G A287141-25G A287141-5G
76988-588EA 431.79 USD
76988-588 76988-590 76988-592 76988-594
(S)-Benzyl 4-isopropyl-2,5-dioxooxazolidine-3-carboxylate 95%
(S)-Benzyl 4-isopropyl-2,5-dioxooxazolidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₅
MW: 277.27 g/mol
MDL Number: MFCD00274015
CAS Number: 158257-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR