4-Chloro-7-ethoxyquinoline 95%

Supplier: Ambeed
Danger

A287111-100MG A287111-250MG
77014-704EA 41 USD
77014-704 77014-706
4-Chloro-7-ethoxyquinoline 95%
4-Chloro-7-ethoxyquinoline

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClNO
MDL Number: MFCD26395197
CAS Number: 178984-50-4
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR