5-Iodothiophene-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A287052-100MG A287052-1G A287052-250MG
77032-230EA 78.51 USD
77032-230 77032-232 77032-234
5-Iodothiophene-2-carboxylic acid 95%
5-Iodothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃IO₂S
MW: 254.05 g/mol
MDL Number: MFCD17215786
CAS Number: 60166-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR