Ethyl 6-bromo-4-oxo-3,4-dihydro-2H-benzo[e][1,3]oxazine-2-carboxylate 97%

Supplier: Ambeed
Warning

A286692-100MG A286692-250MG
77014-700EA 33.95 USD
77014-700 77014-702
Ethyl 6-bromo-4-oxo-3,4-dihydro-2H-benzo[e][1,3]oxazine-2-carboxylate 97%
Ethyl 6-bromo-4-oxo-3,4-dihydro-2H-benzo[e][1,3]oxazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO₄
MW: 300.11 g/mol
MDL Number: MFCD27952666
CAS Number: 30482-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR