(1S,2S)-2-(Diphenylphosphino)cyclohexanamine 98%

Supplier: Ambeed
Warning

A286648-1G A286648-250MG
76988-584EA 497.57 USD
76988-584 76988-586
(1S,2S)-2-(Diphenylphosphino)cyclohexanamine 98%
(1S,2S)-2-(Diphenylphosphino)cyclohexanamine

Precaución: for Research Use Only

Formula: C₁₈H₂₂NP
MW: 283.34 g/mol
MDL Number: MFCD16876055
CAS Number: 452304-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR