5-Fluoropyrimidin-4-amine 97%

Supplier: Ambeed
Warning

A286554-1G A286554-5G A286554-100MG
77032-226EA 70 USD
77032-226 77032-228 77601-262
5-Fluoropyrimidin-4-amine 97%
5-Fluoropyrimidin-4-amine

Precaución: for Research Use Only

Formula: C4H4FN3
MDL Number: MFCD00223685
CAS Number: 811450-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR