Ethyl 2-(benzofuran-3-yl)acetate 97%

Supplier: Ambeed
Warning

A286401-1G A286401-250MG
77032-222EA 130 USD
77032-222 77032-224
Ethyl 2-(benzofuran-3-yl)acetate 97%
Ethyl 2-(benzofuran-3-yl)acetate

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₃
MDL Number: MFCD08060514
CAS Number: 82156-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR