2-Amino-6-azaspiro[3.4]octane-6-carboxylic acid tert-butyl ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 2-amino-6-azaspiro[3.4]octane-6-carboxylate
A286290-1G
A286290-250MG
A286290-100MG
A286290-5G
77014-684EA
314.03
USD
77014-684
77014-686
77612-648
77683-860
2-Amino-6-azaspiro[3.4]octane-6-carboxylic acid tert-butyl ester 98%
2-Amino-6-azaspiro[3.4]octane-6-carboxylic acid tert-butyl ester
Precaución: For research use only.
Formula:
C₁₂H₂₂N₂O₂ MW: 226.32 g/mol |
MDL Number:
MFCD17016203 CAS Number: 1239319-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...