Methyl indoline-6-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A285956-100MG
A285956-10G
A285956-1G
A285956-250MG
A285956-5G
76988-572EA
45.62
USD
76988-572
76988-574
76988-576
76988-578
76988-580
Methyl indoline-6-carboxylate 95%
Methyl indoline-6-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁NO₂ |
MDL Number:
MFCD07371629 CAS Number: 341988-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...