Ethyl 3-(3-chloro-2,4,5-trifluorophenyl)-3-oxopropanoate 97%

Supplier: Ambeed
Warning

A285873-1G A285873-25G A285873-5G
77014-678EA 6.37 USD
77014-678 77014-680 77014-682
Ethyl 3-(3-chloro-2,4,5-trifluorophenyl)-3-oxopropanoate 97%
Ethyl 3-(3-chloro-2,4,5-trifluorophenyl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₁H₈ClF₃O₃
MW: 280.62 g/mol
MDL Number: MFCD09037898
CAS Number: 101987-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR