2,2-Dimethylethenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (2-Methylprop-1-en-1-yl)boronic acid

A285793-100MG A285793-10G A285793-1G A285793-250MG A285793-25G A285793-5G
77014-666EA 5 USD
77014-666 77014-668 77014-670 77014-672 77014-674 77014-676
2,2-Dimethylethenylboronic acid 96%
2,2-Dimethylethenylboronic acid

Precaución: for Research Use Only

Formula: C₄H₉BO₂
MDL Number: MFCD01074609
CAS Number: 14559-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR