2-Chloro-5-iodopyridine 98%
Supplier: Ambeed
|
Warning
|
A285613-100G
A285613-1G
A285613-25G
A285613-500G
A285613-5G
A285613-10G
76988-552EA
146.41
USD
76988-552
76988-554
76988-556
76988-558
76988-560
77683-858
2-Chloro-5-iodopyridine 98%
2-Chloro-5-iodopyridine
Precaución: For research use only.
Formula:
C₅H₃ClIN MW: 239.44 g/mol |
MDL Number:
MFCD01863635 CAS Number: 69045-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...