4-{2-[4-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}morpholine 96%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Morpholinoethoxy)phenylboronic acid, pinacol, 4-(2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)ethyl)morpholine, 4-[2-(Morpholin-4-yl)ethoxy]benzeneboronic acid, pinacol ester

A285332-100MG A285332-1G A285332-250MG A285332-5G
77014-650EA 35.01 USD
77014-650 77014-652 77014-654 77014-656
4-{2-[4-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}morpholine 96%
4-{2-[4-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}morpholine

Precaución: for Research Use Only

Formula: C₁₈H₂₈BNO₄
MW: 333.24 g/mol
MDL Number: MFCD04440857
CAS Number: 690636-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR