N1,N1'-(Ethane-1,2-diyl)bis(N1-methylbenzene-1,4-diamine) 95%
Supplier: Ambeed
|
Warning
|
A285289-100MG
A285289-1G
A285289-250MG
76988-540EA
26.53
USD
76988-540
76988-542
76988-544
N1,N1'-(Ethane-1,2-diyl)bis(N1-methylbenzene-1,4-diamine) 95%
N1,N1'-(Ethane-1,2-diyl)bis(N1-methylbenzene-1,4-diamine)
Precaución: for Research Use Only
Formula:
C₁₆H₂₂N₄ MW: 270.37 g/mol |
MDL Number:
MFCD22376644 CAS Number: 29103-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...