5-Amino-2,4-dihydro-3H-1,2,4-triazol-3-one 98%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1H-1,2,4-triazol-3(2H)-one

A285259-1G A285259-250MG A285259-5G
77032-212EA 180.36 USD
77032-212 77032-214 77032-216
5-Amino-2,4-dihydro-3H-1,2,4-triazol-3-one 98%
5-Amino-2,4-dihydro-3H-1,2,4-triazol-3-one

Precaución: for Research Use Only

Formula: C₂H₄N₄O
MW: 100.079 g/mol
MDL Number: MFCD23140949
CAS Number: 1003-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR