3-Bromo-7-methylimidazo[1,2-a]pyridine-2-carboxylic acid 98+%
Supplier: Ambeed
|
Warning
|
A285146-1G
A285146-100MG
A285146-250MG
76988-534EA
30
USD
76988-534
77890-518
77890-519
3-Bromo-7-methylimidazo[1,2-a]pyridine-2-carboxylic acid 98+%
3-Bromo-7-methylimidazo[1,2-a]pyridine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₇BrN₂O₂ |
MDL Number:
MFCD20921526 CAS Number: 1352723-58-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...