8-Aminoquinolin-6-ol 95%

Supplier: Ambeed
Warning

A284946-100MG A284946-1G A284946-250MG
77032-206EA 35 USD
77032-206 77032-208 77032-210
8-Aminoquinolin-6-ol 95%
8-Aminoquinolin-6-ol

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MDL Number: MFCD06408351
CAS Number: 7402-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR