(2-Ethynyl-5-methoxyphenyl)methanol 95%

Supplier: Ambeed
Warning

A284930-100MG A284930-1G A284930-250MG
77014-638EA 51.99 USD
77014-638 77014-640 77014-642
(2-Ethynyl-5-methoxyphenyl)methanol 95%
(2-Ethynyl-5-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MW: 162.18 g/mol
MDL Number: MFCD27939080
CAS Number: 89414-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR