1'-Methylspiro[indoline-3,4'-piperidine] 95%

Supplier: Ambeed
Warning

A284785-100MG A284785-1G A284785-250MG
77032-200EA 125.19 USD
77032-200 77032-202 77032-204
1'-Methylspiro[indoline-3,4'-piperidine] 95%
1'-Methylspiro[indoline-3,4'-piperidine]

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂
MW: 202.2954 g/mol
MDL Number: MFCD12198567
CAS Number: 69584-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR