(R)-(+)-2,2'-Bis(diphenylphosphino)-6,6'-dimethoxy-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: (R)-MeO-BIPHEP, (R)-(6,6'-Dimethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(diphenylphosphine)

A284492-100MG A284492-250MG A284492-25G A284492-50MG A284492-5G A284492-1G
76988-520EA 18 USD
76988-520 76988-522 76988-524 76988-526 76988-528 77568-574
(R)-(+)-2,2'-Bis(diphenylphosphino)-6,6'-dimethoxy-1,1'-biphenyl 98%
(R)-(+)-2,2'-Bis(diphenylphosphino)-6,6'-dimethoxy-1,1'-biphenyl

Precaución: For research use only.

Formula: C₃₈H₃₂O₂P₂
MDL Number: MFCD03095430
CAS Number: 133545-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR