2,3-Bipyridine-6-carboxylic acid 95%

Supplier: Ambeed
Warning

A284416-250MG A284416-100MG A284416-1G
77014-628EA 143 USD
77014-628 77890-515 77890-516
2,3-Bipyridine-6-carboxylic acid 95%
2,3-Bipyridine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₈N₂O₂
MDL Number: MFCD17017882
CAS Number: 845827-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR