1-(4-Chlorophenyl)cyclopropanamine hydrochloride 98%

Supplier: Ambeed
Warning

A284116-1G A284116-250MG
77032-196EA 53 USD
77032-196 77032-198
1-(4-Chlorophenyl)cyclopropanamine hydrochloride 98%
1-(4-Chlorophenyl)cyclopropanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁Cl₂N
MDL Number: MFCD07995727
CAS Number: 1009102-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR