10-Hydroxydec-2-enoic acid 98%

Supplier: Ambeed
Warning

A283572-100G A283572-1G A283572-25G A283572-5G
77002-232EA 349 USD
77002-232 77002-234 77002-236 77002-238
10-Hydroxydec-2-enoic acid 98%
10-Hydroxydec-2-enoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₃
MDL Number: MFCD00204506
CAS Number: 765-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR