1-(4-Methoxyphenyl)-4-(4-nitrophenyl)piperazine 97%

Supplier: Ambeed
Warning

A283518-100MG A283518-1G A283518-250MG
77032-184EA 39.26 USD
77032-184 77032-186 77032-188
1-(4-Methoxyphenyl)-4-(4-nitrophenyl)piperazine 97%
1-(4-Methoxyphenyl)-4-(4-nitrophenyl)piperazine

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₃O₃
MW: 313.35 g/mol
MDL Number: MFCD01604435
CAS Number: 74852-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR