7-(Benzyloxy)-4-chloro-6-methoxyquinazoline 98%

Supplier: Ambeed
Warning

A283361-10G A283361-1G A283361-250MG A283361-5G
77014-620EA 89.12 USD
77014-620 77014-622 77014-624 77014-626
7-(Benzyloxy)-4-chloro-6-methoxyquinazoline 98%
7-(Benzyloxy)-4-chloro-6-methoxyquinazoline

Precaución: for Research Use Only

Formula: C₁₆H₁₃ClN₂O₂
MW: 300.7396 g/mol
MDL Number: MFCD04115120
CAS Number: 162364-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR