Benzyl-4-(hydroxymethyl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Benzyl 4-(hydroxymethyl)piperidine-1-carboxylate, Benzyl 4-(hydroxymethyl)tetrahydro-1(2H)-pyridinecarboxylate

A283359-100G A283359-10G A283359-25G A283359-500G A283359-5G
77002-218EA 50 USD
77002-218 77002-220 77002-222 77002-224 77002-228 77002-230
Benzyl-4-(hydroxymethyl)piperidine-1-carboxylate 97%
Benzyl-4-(hydroxymethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₃
MDL Number: MFCD02094490
CAS Number: 122860-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR