[4-(Allyloxy)phenyl]boronic acid 98% + stabilised

Supplier: Ambeed
Warning

Synonyms: 4-Allyloxyphenylboronic acid, (4-(Allyloxy)phenyl)boronic acid

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A282814-1G A282814-250MG A282814-5G
77002-206EA 110 USD
77002-206 77002-208 77002-210
[4-(Allyloxy)phenyl]boronic acid 98% + stabilised
[4-(Allyloxy)phenyl]boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₃
MDL Number: MFCD11103728
CAS Number: 1117776-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR