4-(2-Chlorophenyl)-1,3-thiazol-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Chlorophenyl)thiazol-2-amine

A282382-10G A282382-1G A282382-250MG A282382-25G A282382-5G
77002-196EA 171.87 USD
77002-196 77002-198 77002-200 77002-202 77002-204
4-(2-Chlorophenyl)-1,3-thiazol-2-amine 98%
4-(2-Chlorophenyl)-1,3-thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₇ClN₂S
MW: 210.69 g/mol
MDL Number: MFCD01315352
CAS Number: 21344-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR