Fmoc-Lys-OAll·HCl 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Lys-OAll.HCl

A282286-1G A282286-5G A282286-25G
76988-512EA 18.04 USD
76988-512 77612-622 77612-620
Fmoc-Lys-OAll·HCl 97%
Fmoc-Lys-OAll·HCl

Precaución: for Research Use Only

Formula: C₂₄H₂₈N₂O₄·HCl
MW: 444.9 g/mol
MDL Number: MFCD08274506
CAS Number: 815619-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR