L-Threonine benzyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (2S,3R)-Benzyl 2-amino-3-hydroxybutanoate hydrochloride, H-Thr-OBzl.HCl

A282147-100G A282147-1G A282147-250MG A282147-25G A282147-5G
76988-502EA 348 USD
76988-502 76988-504 76988-506 76988-508 76988-510
L-Threonine benzyl ester hydrochloride 98%
L-Threonine benzyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 245.70 g/mol
MDL Number: MFCD00038977
CAS Number: 33645-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR