1-Benzylpiperidine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A281823-1G A281823-250MG A281823-5G
77002-190EA 103.97 USD
77002-190 77002-192 77002-194
1-Benzylpiperidine-3-carboxylic acid 95%
1-Benzylpiperidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₂
MW: 219.28 g/mol
MDL Number: MFCD03086172
CAS Number: 141943-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR