tert-Butyl (2-cyanopyridin-4-yl)carbamate 95%

Supplier: Ambeed
Warning

A281736-100MG A281736-1G A281736-250MG
77032-172EA 25 USD
77032-172 77032-174 77032-176
tert-Butyl (2-cyanopyridin-4-yl)carbamate 95%
tert-Butyl (2-cyanopyridin-4-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃O₂
MW: 219.24 g/mol
MDL Number: MFCD11975697
CAS Number: 262295-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR