3,5-Dibromopyridine-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibromoisonicotinic acid

A281379-1G A281379-25G A281379-5G
77032-166EA 13.8 USD
77032-166 77032-168 77032-170
3,5-Dibromopyridine-4-carboxylic acid 98%
3,5-Dibromopyridine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃Br₂NO₂
MW: 280.91 g/mol
MDL Number: MFCD08689615
CAS Number: 13958-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR